2-Amino-5-fluoropyridine-3-carbaldehyde

Supplier: Matrix Scientific

055570-500MG
101909-696EA 730.77 USD
101909-696
2-Amino-5-fluoropyridine-3-carbaldehyde
2-Amino-5-fluoropyridine-3-carbaldehyde

Hazard: Irritant

Formula: C₆H₅FN₂O
MDL Number: MFCD13190372
CAS Number: 1188433-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR