5-Fluoro-3-(3-hydroxypropyl)pyridin-2-ol
Supplier: Matrix Scientific
055562-100MG
101909-680EA
364.62
USD
101909-680
5-Fluoro-3-(3-hydroxypropyl)pyridin-2-ol
5-Fluoro-3-(3-hydroxypropyl)pyridin-2-ol
Hazard: Irritant
Formula:
C₈H₁₀FNO₂ |
MDL Number:
MFCD16628226 CAS Number: 1228666-37-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...