5-Fluoro-2-pivalamidonicotinic acid

Supplier: Matrix Scientific

055558-100MG
101909-672EA 364.62 USD
101909-672
5-Fluoro-2-pivalamidonicotinic acid
5-Fluoro-2-pivalamidonicotinic acid

Hazard: Irritant

Formula: C₁₁H₁₃FN₂O₃
MDL Number: MFCD16628223
CAS Number: 1228666-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR