(trans)-1-tert-Butyl-3-methyl-4-(2-fluoro-6-(3-(hydroxymethyl)pyrrolidin-1-yl)pyridin-3-yl)pyrrolidine-1,3-dicarboxylate

Supplier: Matrix Scientific

055551-100MG
101909-658EA 364.62 USD
101909-658
(trans)-1-tert-Butyl-3-methyl-4-(2-fluoro-6-(3-(hydroxymethyl)pyrrolidin-1-yl)pyridin-3-yl)pyrrolidine-1,3-dicarboxylate
(trans)-1-tert-Butyl-3-methyl-4-(2-fluoro-6-(3-(hydroxymethyl)pyrrolidin-1-yl)pyridin-3-yl)pyrrolidine-1,3-dicarboxylate

Hazard: Irritant

Formula: C₂₁H₃₀FN₃O₅
MDL Number: MFCD16628216
CAS Number: 1228663-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR