N-(5-Fluoro-3-iodopyridin-2-yl)pivalamide

Supplier: Matrix Scientific

055548-100MG
101909-652EA 364.62 USD
101909-652
N-(5-Fluoro-3-iodopyridin-2-yl)pivalamide
N-(5-Fluoro-3-iodopyridin-2-yl)pivalamide

Hazard: Irritant

Formula: C₁₀H₁₂FIN₂O
MDL Number: MFCD16628213
CAS Number: 823218-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR