2-Fluoro-6-(pyrrolidin-1-yl)isonicotinonitrile

Supplier: Matrix Scientific

055547-100MG
101909-650EA 364.62 USD
101909-650
2-Fluoro-6-(pyrrolidin-1-yl)isonicotinonitrile
2-Fluoro-6-(pyrrolidin-1-yl)isonicotinonitrile

Hazard: Irritant

Formula: C₁₀H₁₀FN₃
MDL Number: MFCD15530272
CAS Number: 1228666-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR