(E)-Methyl-3-(2-fluoro-6-(pyrrolidin-1-yl)pyridin-4-yl)acrylate
Supplier: Matrix Scientific
055542-100MG
101909-640EA
364.62
USD
101909-640
(E)-Methyl-3-(2-fluoro-6-(pyrrolidin-1-yl)pyridin-4-yl)acrylate
(E)-Methyl-3-(2-fluoro-6-(pyrrolidin-1-yl)pyridin-4-yl)acrylate
Hazard: Irritant
Formula:
C₁₃H₁₅FN₂O₂ |
MDL Number:
MFCD15530268 CAS Number: 1228670-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...