5-Bromo-2-pivalamidopyridin-4-yl pivalate

Supplier: Matrix Scientific

055541-100MG
101909-638EA 364.62 USD
101909-638
5-Bromo-2-pivalamidopyridin-4-yl pivalate
5-Bromo-2-pivalamidopyridin-4-yl pivalate

Hazard: Irritant

Formula: C₁₅H₂₁BrN₂O₃
MDL Number: MFCD15530267
CAS Number: 1228666-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR