(6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoropyridin-3-yl)methanol

Supplier: Matrix Scientific

055529-100MG
101909-614EA 364.62 USD
101909-614
(6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoropyridin-3-yl)methanol
(6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoropyridin-3-yl)methanol

Hazard: Irritant

Formula: C₁₇H₂₉FN₂O₂Si
MDL Number: MFCD15530255
CAS Number: 1228665-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR