(E)-6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoronicotinaldehyde oxime

Supplier: Matrix Scientific

055528-100MG
101909-612EA 364.62 USD
101909-612
(E)-6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoronicotinaldehyde oxime
(E)-6-(3-((tert-Butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoronicotinaldehyde oxime

Hazard: Irritant

Formula: C₁₇H₂₈FN₃O₂Si
MDL Number: MFCD15530254
CAS Number: 1228670-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR