(E)-Methyl-3-(6-(3-((tert-butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoropyridin-3-yl)acrylate

Supplier: Matrix Scientific

055525-100MG
101909-606EA 364.62 USD
101909-606
(E)-Methyl-3-(6-(3-((tert-butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoropyridin-3-yl)acrylate
(E)-Methyl-3-(6-(3-((tert-butyldimethylsilyloxy)methyl)pyrrolidin-1-yl)-2-fluoropyridin-3-yl)acrylate

Hazard: Irritant

Formula: C₂₀H₃₁FN₂O₃Si
MDL Number: MFCD15530251
CAS Number: 1228670-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR