(E)-Methyl-3-(2-methoxy-6-(pyrrolidin-1-yl)-pyridin-3-yl)acrylate

Supplier: Matrix Scientific

055522-100MG
101909-600EA 364.62 USD
101909-600
(E)-Methyl-3-(2-methoxy-6-(pyrrolidin-1-yl)-pyridin-3-yl)acrylate
(E)-Methyl-3-(2-methoxy-6-(pyrrolidin-1-yl)-pyridin-3-yl)acrylate

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₃
MDL Number: MFCD15530248
CAS Number: 1228670-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR