8-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

055517-100MG
101909-590EA 406.35 USD
101909-590
8-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro[1,4]dioxino[2,3-b]pyridine
8-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₃H₁₈BNO₄
MW: 263.1 g/mol
MDL Number: MFCD16628208
CAS Number: 1309980-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR