7-Iodo-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Supplier: Matrix Scientific

055515-100MG
101909-586EA 246.15 USD
101909-586
7-Iodo-2,3-dihydro[1,4]dioxino[2,3-b]pyridine
7-Iodo-2,3-dihydro[1,4]dioxino[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₆INO₂
MW: 263.04 g/mol
MDL Number: MFCD16628206
CAS Number: 1228666-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR