6-Fluoro-3,4-dihydro-2H-pyrano[2,3-b]pyridine

Supplier: Matrix Scientific

055512-100MG
101909-580EA 364.62 USD
101909-580
6-Fluoro-3,4-dihydro-2H-pyrano[2,3-b]pyridine
6-Fluoro-3,4-dihydro-2H-pyrano[2,3-b]pyridine

Hazard: Irritant

Formula: C₈H₈FNO
MDL Number: MFCD16628204
CAS Number: 1228666-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR