6,8-Diiodo-3,4-dihydro-2H-pyrano[3,2-b]pyridine

Supplier: Matrix Scientific

055511-100MG
101909-578EA 364.62 USD
101909-578
6,8-Diiodo-3,4-dihydro-2H-pyrano[3,2-b]pyridine
6,8-Diiodo-3,4-dihydro-2H-pyrano[3,2-b]pyridine

Hazard: Irritant

Formula: C₈H₇I₂NO
MDL Number: MFCD16628203
CAS Number: 1222533-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR