3,4-Dihydro-2H-pyrano[2,3-b]pyridine-6-carbaldehyde

Supplier: Matrix Scientific

055509-100MG
101909-574EA 364.62 USD
101909-574
3,4-Dihydro-2H-pyrano[2,3-b]pyridine-6-carbaldehyde
3,4-Dihydro-2H-pyrano[2,3-b]pyridine-6-carbaldehyde

Hazard: Irritant

Formula: C₉H₉NO₂
MDL Number: MFCD16628201
CAS Number: 1222533-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR