tert-Butyl-1,9-diazaspiro[5.5]undecane-9-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1,9-Diazaspiro[5.5]undecane, N9-BOC protected

055461-500MG
101909-384EA 1016.92 USD
101909-384
tert-Butyl-1,9-diazaspiro[5.5]undecane-9-carboxylate ≥95%
tert-Butyl-1,9-diazaspiro[5.5]undecane-9-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₆N₂O₂
MW: 254.38 g/mol
MDL Number: MFCD09998745
CAS Number: 1031927-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR