4-[(tert-Butoxycarbonyl)amino]-2-(4-methylbenzyl)butanoic acid ≥95%

Supplier: Matrix Scientific

055454-500MG
101909-362EA 556.92 USD
101909-362
4-[(tert-Butoxycarbonyl)amino]-2-(4-methylbenzyl)butanoic acid ≥95%
4-[(tert-Butoxycarbonyl)amino]-2-(4-methylbenzyl)butanoic acid

Hazard: Irritant

Formula: C₁₇H₂₅NO₄
MDL Number: MFCD16622775
CAS Number: 853680-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR