Tetrakis(4-aminophenyl)methane ≥98%

Supplier: Matrix Scientific

Synonyms: 4,4',4'',4'''-Methanetetrayltetraaniline

055403-500MG
101909-188EA 300 USD
101909-188
Tetrakis(4-aminophenyl)methane ≥98%
Tetrakis(4-aminophenyl)methane

Hazard: Irritant

Formula: C₂₅H₂₄N₄
MW: 380.5 g/mol
Melting Pt: >300 °C
MDL Number: MFCD12546930
CAS Number: 60532-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR