2-Cyano-3,6-dichloropyridine ≥98%
Supplier: Matrix Scientific
Synonyms:
3,6-Dichloropicolinonitrile
055400-5G
055400-1G
101909-178EA
69.23
USD
101909-178
101909-180
2-Cyano-3,6-dichloropyridine ≥98%
2-Cyano-3,6-dichloropyridine
Hazard: Irritant
Formula:
C₆H₂Cl₂N₂ MW: 173 g/mol |
MDL Number:
MFCD00546824 CAS Number: 1702-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...