3,4-Dichloro-2,5,6-trifluoropyridine ≥98%

Supplier: Matrix Scientific

054969-1G 054969-5G
101909-170EA 181.54 USD
101909-170 101909-172
3,4-Dichloro-2,5,6-trifluoropyridine ≥98%
3,4-Dichloro-2,5,6-trifluoropyridine

Hazard: Irritant

Formula: C₅Cl₂F₃N
MDL Number: MFCD12024847
CAS Number: 52026-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR