4-(Difluoromethyl)-1-fluoro-2-methylbenzene ≥95%

Supplier: Matrix Scientific

054964-1G
101909-162EA 200 USD
101909-162
4-(Difluoromethyl)-1-fluoro-2-methylbenzene ≥95%
4-(Difluoromethyl)-1-fluoro-2-methylbenzene

Hazard: Irritant

Formula: C₈H₇F₃
MDL Number: MFCD14698601
CAS Number: 1214372-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR