1-Chloro-4-(difluoromethyl)-2-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

054962-1G
101909-158EA 320 USD
101909-158
1-Chloro-4-(difluoromethyl)-2-(trifluoromethyl)benzene ≥95%
1-Chloro-4-(difluoromethyl)-2-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₄ClF₅
MDL Number: MFCD14698557
CAS Number: 1214338-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR