4-Chloro-1-(difluoromethyl)-2-fluorobenzene ≥95%

Supplier: Matrix Scientific

054957-1G
101909-148EA 158.46 USD
101909-148
4-Chloro-1-(difluoromethyl)-2-fluorobenzene ≥95%
4-Chloro-1-(difluoromethyl)-2-fluorobenzene

Hazard: Irritant

Formula: C₇H₄ClF₃
MDL Number: MFCD14698547
CAS Number: 1214337-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR