1-Bromo-2-(difluoromethyl)-4-fluorobenzene ≥95%

Supplier: Matrix Scientific

054951-1G
101909-136EA 186.15 USD
101909-136
1-Bromo-2-(difluoromethyl)-4-fluorobenzene ≥95%
1-Bromo-2-(difluoromethyl)-4-fluorobenzene

Hazard: Irritant

Formula: C₇H₄BrF₃
MDL Number: MFCD14698532
CAS Number: 1198171-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR