2,6-Dichloro-3-nitrobenzotrifluoride ≥95%

Supplier: Matrix Scientific

054946-1G
101909-132EA 375.38 USD
101909-132
2,6-Dichloro-3-nitrobenzotrifluoride ≥95%
2,6-Dichloro-3-nitrobenzotrifluoride

Hazard: Irritant-Harmful/Moisture Sensitive

Formula: C₇H₂Cl₂F₃NO₂
MDL Number: MFCD16140179
CAS Number: 1221272-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR