3-Methyl-2-nitrobenzotrifluoride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-2-nitro-3-(trifluoromethyl)benzene

054939-1G
101909-118EA 79.35 USD
101909-118
3-Methyl-2-nitrobenzotrifluoride ≥95%
3-Methyl-2-nitrobenzotrifluoride

Hazard: Irritant

Formula: C₈H₆F₃NO₂
CAS Number: 92891-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR