1-(Difluoromethyl)naphthalene ≥97%

Supplier: Matrix Scientific

054937-1G
101909-114EA 178.46 USD
101909-114
1-(Difluoromethyl)naphthalene ≥97%
1-(Difluoromethyl)naphthalene

Hazard: Irritant

Formula: C₁₁H₈F₂
MDL Number: MFCD12546939
CAS Number: 53731-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR