1-Nitro-2-(2,2,2-trifluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

054933-1G
101909-106EA 75.38 USD
101909-106
1-Nitro-2-(2,2,2-trifluoroethyl)benzene ≥95%
1-Nitro-2-(2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₆F₃NO₂
MDL Number: MFCD14525517
CAS Number: 145914-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR