1-Methyl-4-(2,2,2-trifluoro-1-methylethyl)benzene ≥95%

Supplier: Matrix Scientific

054932-1G
101909-104EA 315.38 USD
101909-104
1-Methyl-4-(2,2,2-trifluoro-1-methylethyl)benzene ≥95%
1-Methyl-4-(2,2,2-trifluoro-1-methylethyl)benzene

Hazard: Irritant

Formula: C₁₀H₁₁F₃
MDL Number: MFCD14525514
CAS Number: 1204295-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR