2,5-Dichloro-1-nitro-3-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

054931-1G
101909-102EA 364.62 USD
101909-102
2,5-Dichloro-1-nitro-3-(trifluoromethyl)benzene ≥95%
2,5-Dichloro-1-nitro-3-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₇H₂Cl₂F₃NO₂
MDL Number: MFCD00047673
CAS Number: 98373-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR