1,1,1-Trifluoro-2,2-diphenylpropane ≥95%

Supplier: Matrix Scientific

054929-1G
101909-098EA 250.77 USD
101909-098
1,1,1-Trifluoro-2,2-diphenylpropane ≥95%
1,1,1-Trifluoro-2,2-diphenylpropane

Hazard: Irritant

Formula: C₁₅H₁₃F₃
MDL Number: MFCD14525526
CAS Number: 112754-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR