trans-1-n-Pentyl-4-(trifluoromethyl)cyclohexane ≥97%

Supplier: Matrix Scientific

054928-1G
101909-096EA 267.69 USD
101909-096
trans-1-n-Pentyl-4-(trifluoromethyl)cyclohexane ≥97%
trans-1-n-Pentyl-4-(trifluoromethyl)cyclohexane

Hazard: Irritant

Formula: C₁₂H₂₁F₃
MDL Number: MFCD11226629
CAS Number: 1212501-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR