3-Chloro-2,6-difluoropyridine ≥98%

Supplier: Matrix Scientific

054926-1G 054926-5G
101909-088EA 73.85 USD
101909-088 101909-090
3-Chloro-2,6-difluoropyridine ≥98%
3-Chloro-2,6-difluoropyridine

Hazard: Flammable/Corrosive

Formula: C₅H₂ClF₂N
MDL Number: MFCD11044285
CAS Number: 52208-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR