2-Phenethylpyrazine ≥95%

Supplier: Matrix Scientific

054915-500MG 054915-1G
101909-042EA 356.92 USD
101909-042 101909-040
2-Phenethylpyrazine ≥95%
2-Phenethylpyrazine

Hazard: Irritant

Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
MDL Number: MFCD16140369
CAS Number: 91391-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR