N-Allyl-3-amino-4-chlorobenzenesulfonamide

Supplier: Matrix Scientific

052835-2.5G 052835-500MG
101901-228EA 364.62 USD
101901-228 101901-230
N-Allyl-3-amino-4-chlorobenzenesulfonamide
N-Allyl-3-amino-4-chlorobenzenesulfonamide

Hazard: Irritant

Formula: C₉H₁₁ClN₂O₂S
MDL Number: MFCD12791542
CAS Number: 1220034-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR