N-Allyl-2-amino-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

052827-500MG 052827-2.5G
101901-196EA 364.62 USD
101901-196 101901-198
N-Allyl-2-amino-3-methylbutanamide hydrochloride
N-Allyl-2-amino-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O
MDL Number: MFCD13562396
CAS Number: 1246172-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR