N-Allyl-3-aminopropanamide hydrochloride
Supplier: Matrix Scientific
052826-2.5G
052826-500MG
101901-192EA
364.62
USD
101901-192
101901-194
N-Allyl-3-aminopropanamide hydrochloride
N-Allyl-3-aminopropanamide hydrochloride
Hazard: Irritant
Formula:
C₆H₁₃ClN₂O |
MDL Number:
MFCD13562395 CAS Number: 1220034-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...