N-Allyl-5-bromo-2-pyridinamine
Supplier: Matrix Scientific
052821-2.5G
052821-500MG
101901-172EA
364.62
USD
101901-172
101901-174
N-Allyl-5-bromo-2-pyridinamine
N-Allyl-5-bromo-2-pyridinamine
Hazard: Irritant
Formula:
C₈H₉BrN₂ |
MDL Number:
MFCD13562390 CAS Number: 106037-57-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...