N-Allyl-2-chloronicotinamide

Supplier: Matrix Scientific

052819-500MG 052819-2.5G
101901-164EA 364.62 USD
101901-164 101901-166
N-Allyl-2-chloronicotinamide
N-Allyl-2-chloronicotinamide

Hazard: Irritant

Formula: C₉H₉ClN₂O
MDL Number: MFCD03389949
CAS Number: 545372-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR