2-(Allylamino)isonicotinic acid

Supplier: Matrix Scientific

052816-2.5G 052816-500MG
101901-152EA 166.15 USD
101901-152 101901-154
2-(Allylamino)isonicotinic acid
2-(Allylamino)isonicotinic acid

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
MDL Number: MFCD11131942
CAS Number: 1019461-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR