Ethyl-4-(allylamino)-3-aminobenzoate
Supplier: Matrix Scientific
052811-2.5G
052811-500MG
101901-132EA
364.62
USD
101901-132
101901-134
Ethyl-4-(allylamino)-3-aminobenzoate
Ethyl-4-(allylamino)-3-aminobenzoate
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₂ |
MDL Number:
MFCD13562385 CAS Number: 1220029-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...