N-Butyl-4-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

Synonyms: N-Butylpiperidine-4-carboxamide

052808-500MG 052808-2.5G
101901-120EA 364.62 USD
101901-120 101901-122
N-Butyl-4-piperidinecarboxamide hydrochloride
N-Butyl-4-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O
MW: 220.74 g/mol
MDL Number: MFCD11615740
CAS Number: 73415-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR