2-(Butylamino)-5-bromopyridine

Supplier: Matrix Scientific

052796-500MG 052796-2.5G
101901-072EA 364.62 USD
101901-072 101901-074
2-(Butylamino)-5-bromopyridine
2-(Butylamino)-5-bromopyridine

Hazard: Irritant

Formula: C₉H₁₃BrN₂
MDL Number: MFCD11123176
CAS Number: 280116-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR