3-Amino-4-chloro-N-propylbenzenesulfonamide

Supplier: Matrix Scientific

052784-500MG 052784-2.5G
101901-024EA 364.62 USD
101901-024 101901-026
3-Amino-4-chloro-N-propylbenzenesulfonamide
3-Amino-4-chloro-N-propylbenzenesulfonamide

Hazard: Irritant

Formula: C₉H₁₃ClN₂O₂S
MDL Number: MFCD09900720
CAS Number: 1017409-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR