2-Amino-3-phenyl-N-propylpropanamide hydrochloride

Supplier: Matrix Scientific

052778-500MG 052778-2.5G
101901-000EA 364.62 USD
101901-000 101901-002
2-Amino-3-phenyl-N-propylpropanamide hydrochloride
2-Amino-3-phenyl-N-propylpropanamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂O
MDL Number: MFCD13562365
CAS Number: 934505-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR