2-(Ethylamino)nicotinic acid

Supplier: Matrix Scientific

052726-2.5G 052726-500MG
101900-792EA 994.64 USD
101900-792 101900-790
2-(Ethylamino)nicotinic acid
2-(Ethylamino)nicotinic acid

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD00128790
CAS Number: 669087-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR