3-Chloro-N-ethyl-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

052723-500MG 052723-2.5G
101900-778EA 364.62 USD
101900-778 101900-780
3-Chloro-N-ethyl-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-ethyl-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₈H₈ClF₃N₂
MW: 224.61 g/mol
MDL Number: MFCD11122453
CAS Number: 1042499-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR