2-Amino-N-2-dimethylpropanamide hydrochloride
Supplier: Matrix Scientific
052711-500MG
052711-2.5G
101900-732EA
838.46
USD
101900-732
101900-730
2-Amino-N-2-dimethylpropanamide hydrochloride
2-Amino-N-2-dimethylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₅H₁₃ClN₂O |
MDL Number:
MFCD13562339 CAS Number: 84827-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...